EMAIL THIS PAGE TO A FRIEND
05661501

Neroli oil

secondary reference standard

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Browse by Matrix, Browse by certified Analyte or Parameter, Essential oils,
CofA   certificate is enclosed in each package
shelf life   limited shelflife, expiry date on the label
mfr. no.   HWI
Featured Industry   Cleaning Products
Cosmetics
Flavors and Fragrances
Food and Beverages
Personal Care
storage temp.   room temp

Description

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phytochemicals: Linalool (quantitative); Limonene, β-Pinene, Geranyl acetate (qualitative)

Safety & Documentation

Safety Information

WGK Germany 
WGK 2
RTECS 
QQ0960000
Flash Point(F) 
158.0 °F - closed cup
Flash Point(C) 
70 °C - closed cup

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?