EMAIL THIS PAGE TO A FRIEND
05961501

Spruce needle oil

secondary reference standard

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Essential oils, Flavor and Fragrance Standards, Flavors and Fragrance Standards,
CofA   certificate is enclosed in each package
shelf life   limited shelf life, expiry date on the label
Featured Industry   Cleaning Products
Cosmetics
Food and Beverages
Personal Care

Description

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phytochemicals: (-)-Bornyl acetate (quantitative); α-Pinene, Camphene, (+)-3-d-Caren, Limonene (qualitative)

Safety & Documentation

Safety Information

RIDADR 
UN 1993C 3 / PGIII

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?