• USA Home
  • 06040 - QPCRScan Listeria spp.

EMAIL THIS PAGE TO A FRIEND
06040

QPCRScan Listeria spp.

for microbiology

  •  NACRES NA.85

Purchase

Properties

Related Categories Identification Tests & Reagents, Microbiology, Molecular Biological, qPCR Kits
Quality Level   100
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
application(s)   microbiology: suitable (microbial identification)
Featured Industry   Environmental
Food and Beverages
suitability   Listeria spp.

Description

Application

Detection of Listeria spp. by Real time PCR.

Components

Content for 96 reactions:
qPCR Master-Mix Probe, lyophilized (for 1 ml, in Resuspension Buffer Probe; Cat. No. 91017)
Primer/Probe Mix lyophilized (for 110 μl, in water, Cat. No. 92484)
H2O (PCR grade) 2 ml (Cat. No. 90613)
Resuspension Buffer Probe 1.1 ml
Positive Control DNA lyophilized (for 1.25 ml, in water; Cat. No. 90616)

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?