• USA Home
  • 06731 - (R)-Boc-2-methyl-β-Phe-OH

EMAIL THIS PAGE TO A FRIEND
06731 Sigma-Aldrich

(R)-Boc-2-methyl-β-Phe-OH

≥98.0%

Synonym: (R)-3-(Boc-amino)-3-(2-methylphenyl)propionic acid, Boc-2-methyl-L-β-phenylalanine

Purchase

Properties

Related Categories Beta-Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
assay   ≥98.0%
form   powder
application(s)   peptide synthesis: suitable
color   white
mp   165-166 °C
storage temp.   room temp
SMILES string   Cc1ccccc1[C@@H](CC(O)=O)NC(=O)OC(C)(C)C
InChI   1S/C15H21NO4/c1-10-7-5-6-8-11(10)12(9-13(17)18)16-14(19)20-15(2,3)4/h5-8,12H,9H2,1-4H3,(H,16,19)(H,17,18)/t12-/m1/s1
InChI key   WWDMSBGBNGEQDH-GFCCVEGCSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

β-Amino Acids and Homologs

Although they are less abundant than their α-analogues, β-amino acids occur in nature both in free form and bound to peptides. Oligomers composed exclusively of β-amino acids (so-called β-peptides) m...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 11.
Keywords: Chemfiles, Degradations, Metabolism, Peptidomimetics

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?