EMAIL THIS PAGE TO A FRIEND
07406 Supelco

m-Anisaldehyde

analytical standard

Synonym: 3-Methoxybenzaldehyde

  • CAS Number 591-31-1

  • Linear Formula CH3OC6H4CHO

  • Molecular Weight 136.15

  •  Beilstein/REAXYS Number 606013

  •  EC Number 209-712-8

  •  MDL number MFCD00003361

  •  PubChem Substance ID 329748429

  •  NACRES NA.24

Purchase

Properties

Related Categories A-F, A-L, Aldehyde, All Alphabetically Sorted, Analytical Standards,
Quality Level   100
grade   analytical standard
assay   ≥98.0% (GC)
shelf life   limited shelf life, expiry date on the label
refractive index   n20/D 1.553 (lit.)
bp   143 °C/50 mmHg (lit.)
density   1.117 g/mL at 20 °C (lit.)
Featured Industry   Cleaning Products
Cosmetics
Flavors and Fragrances
Food and Beverages
Personal Care
SMILES string   [H]C(=O)c1cccc(OC)c1
InChI   1S/C8H8O2/c1-10-8-4-2-3-7(5-8)6-9/h2-6H,1H3
InChI key   WMPDAIZRQDCGFH-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
BZ2605000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?