EMAIL THIS PAGE TO A FRIEND
07507 Sigma-Aldrich

Tryptone Water

for microbiology, ready to use media in the bottle

Purchase

Properties

Related Categories All Food Control Media (alphabetical), All Liquid Media/Broths, Bacteria (General Media), Biochemical Identification Media by Test, Biochemical Identification Media, A to Z,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   sodium chloride, 5.0 g/L
  tryptone (tryptic hydrolysate of casein), 10.0 g/L
final pH   7.3±0.2 (25 °C)
Featured Industry   Environmental
Food and Beverages
suitability   nonselective for Bacteria (General Media)
  nonselective for Enterobacter
  nonselective for Enterobacteriaceae
  nonselective for Escherichia coli and Coliforms
  nonselective for Proteus
  nonselective for Salmonella
  nonselective for Staphylococcus

Description

Application

Substrate with low nutrient capacity, for the research of indole production in coliform microorganisms according ISO 7251 standard.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

EZ-Product Family for Bioburden Testing

The EZ family has been designed to optimize and streamline your bioburden workflow. Ideal for microbiological water, beverage, and medical device testing using the membrane filtration method. The fam...
Keywords: Filtration

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the natural environment, microbes have adapted to the habitats most suitable ...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?