• USA Home
  • 07883 - Hexadecyltrimethylammonium bromide

EMAIL THIS PAGE TO A FRIEND
07883

Hexadecyltrimethylammonium bromide

puriss. p.a., matrix substance for MALDI-MS

Synonym: CTAB, Cetrimonium bromide, Cetyltrimethylammonium bromide, Palmityltrimethylammonium bromide

  • CAS Number 57-09-0

  • Linear Formula CH3(CH2)15N(Br)(CH3)3

  • Molecular Weight 364.45

  •  Beilstein/REAXYS Number 3598189

  •  EC Number 200-311-3

  •  MDL number MFCD00011772

  •  PubChem Substance ID 329748525

Purchase

Properties

grade   matrix substance for MALDI-MS
  puriss. p.a.
form   solid
pH   5.0-7 (25 °C, 36.4 g/L)
mp   248-251 °C (lit.)
solubility   H2O: soluble 36.4 g/L at 20 °C (completely)
SMILES string   [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
InChI   1S/C19H42N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H/q+1;/p-1
InChI key   LZZYPRNAOMGNLH-UHFFFAOYSA-M
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Target organs 
Gastrointestinal tract
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
Flash Point(F) 
471.2 °F
Flash Point(C) 
244 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?