• USA Home
  • 09587 - Grubbs Catalyst 1st Generation

EMAIL THIS PAGE TO A FRIEND
09587 Sigma-Aldrich

Grubbs Catalyst 1st Generation

purum, catalyst grade (for olefin metathesis), ≥97.0% (C)

Synonym: Benzylidene-bis(tricyclohexylphosphine)dichlororuthenium, Bis(tricyclohexylphosphine)benzylidine ruthenium(IV) dichloride, Dichloro(benzylidene)bis(tricyclohexylphosphine)ruthenium(II), Grubbs Catalyst C823

  • CAS Number 172222-30-9

  • Empirical Formula (Hill Notation) C43H72Cl2P2Ru

  • Molecular Weight 822.96

  •  MDL number MFCD01090946

Purchase

Properties

grade   catalyst grade (for olefin metathesis)
InChI Key   IEEOQDKBWLBRKR-UHFFFAOYSA-L
grade   purum
assay   ≥97.0% (C)
mp   153 °C (dec.) (lit.)
storage temp.   2-8°C

Description

Other Notes

Highly efficient catalyst for various types of metathesis reactions

Legal Information

Grubbs Catalyst is a trademark of Umicore

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

579726 Grubbs Catalyst 1st Generation, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?