EMAIL THIS PAGE TO A FRIEND
09688 Sigma-Aldrich

Ammonium acetate

BioUltra, for molecular biology, ≥99.0% (calc. based on dry substance, T)

  • CAS Number 631-61-8

  • Linear Formula CH3CO2NH4

  • Molecular Weight 77.08

  •  Beilstein/REAXYS Number 4186741

  •  EC Number 211-162-9

  •  MDL number MFCD00013066

  •  eCl@ss 39021908

  •  PubChem Substance ID 329748799

Purchase

Properties

grade   for molecular biology
product line   BioUltra
assay   ≥99.0% (calc. based on dry substance, T)
form   solid
impurities   DNases, none detected
  RNases, none detected
  insoluble matter, passes filter test
  phosphatases, none detected
  proteases, none detected
  ≤2% water
ign. residue   ≤0.01% (as SO4)
pH   6.5-7.5 (25 °C, 1 M in H2O)
mp   110-112 °C (dec.) (lit.)
solubility   H2O: 1 M at 20 °C, clear, colorless
anion traces   chloride (Cl-): ≤5 mg/kg
  nitrate (NO3-): ≤10 mg/kg
  sulfate (SO42-): ≤10 mg/kg
cation traces   Al: ≤5 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤2 mg/kg
  K: ≤50 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Zn: ≤5 mg/kg
λ   1 M in H2O
SMILES string   N.CC(O)=O
UV absorption   λ: 260 nm Amax: ≤0.015
  λ: 280 nm Amax: ≤0.010
InChI   1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3
InChI key   USFZMSVCRYTOJT-UHFFFAOYSA-N

Description

Application

Ammonium acetate can be used to study molecular biology, reagents, protein structural analysis, proteomics and x-ray crystallography. Ammonium acetate has been used in a study to develop an easy, rapid, low-cost and effective cetyltrimethylammonium bromide (CTAB)-ammonium acetate method for high-quality RNA isolation from polysaccharide- and polyphenol-rich cotton tissues. Ammonium acetate has also been used in a study to develop and validate a simple and rapid high-performance liquid chromatography-electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) method for the determination of 4-p-anisamidobutyric acid (ABA; or N-anysoyl-γ-aminobutiryc acid, N-anisoyl-GABA), a major active metabolite of aniracetam, in human plasma.

Other Notes

Alternative to sodium acetate in ethanol precipitation of nucleic acids. Prevents precipitation of deoxyribonucleoside triphosphates.; Useful for removal of untreated triphosphates from the products of reverse transcriptase, DNA polymerase or terminal polymerase-catalyzed reactions.

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
1
RTECS 
AF3675000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?