• USA Home
  • 10605 - 9-Anthracenecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
10605 Sigma-Aldrich

9-Anthracenecarboxylic acid

purum, for fluorescence, ≥96.0% (T)

Synonym: 9-Anthroic acid

  • CAS Number 723-62-6

  • Empirical Formula (Hill Notation) C15H10O2

  • Molecular Weight 222.24

  •  Beilstein/REAXYS Number 1875336

  •  EC Number 211-964-9

  •  MDL number MFCD00001257

  •  PubChem Substance ID 329748909

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Biochemicals and Reagents, Building Blocks, C13 to C42+,
grade   for fluorescence
  purum
assay   ≥96.0% (T)
impurities   ≤1% anthracene
ign. residue   ≤0.5%
mp   213-217 °C (lit.)
solubility   H2O: soluble
  ethanol: soluble
fluorescence   λex 255 nm; λem 411 nm in ethanol
SMILES string   OC(=O)c1c2ccccc2cc3ccccc13
InChI   1S/C15H10O2/c16-15(17)14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)14/h1-9H,(H,16,17)
InChI key   XGWFJBFNAQHLEF-UHFFFAOYSA-N

Description

Analysis Note

In addition to the emission maximum at 411 nm, there are lower maxima at 388, 435 and 458 nm.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A89405 9-Anthracenecarboxylic acid, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?