• USA Home
  • 106380 - 3-Amino-9-ethylcarbazole

EMAIL THIS PAGE TO A FRIEND
106380 Sigma-Aldrich

3-Amino-9-ethylcarbazole

technical grade, 90%

Synonym: 9-Ethylcarbazol-3-amine, AEC

  • CAS Number 132-32-1

  • Empirical Formula (Hill Notation) C14H14N2

  • Molecular Weight 210.27

  •  Beilstein/REAXYS Number 174969

  •  EC Number 205-057-7

  •  MDL number MFCD00004964

Purchase

Properties

grade   technical grade
InChI Key   OXEUETBFKVCRNP-UHFFFAOYSA-N
assay   90%
impurities   <3% ethanol
  3% solvent crystallizaion

Description

Specificity

3-Amino-9-ethylcarbazole is a peroxidase substrate suitable for use in immunoblotting and immunohistochemical staining procedures. This substrate produces an insoluble end product that is red in color and can be observed visually.

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?