EMAIL THIS PAGE TO A FRIEND
11099 Sigma-Aldrich

Arsenic(III) oxide

puriss. p.a., ACS reagent, 99.95-100.05% (RT)

Synonym: Arsenic trioxide, Arsenous acid

Purchase

Properties

Related Categories A-B, Puriss p.a. ACS, Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Arsenic,
grade   ACS reagent
  puriss. p.a.
assay   99.95-100.05% (RT)
SMILES string   O=[As]O[As]=O
InChI   1S/As2O3/c3-1-5-2-4
InChI key   IKWTVSLWAPBBKU-UHFFFAOYSA-N

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN 1561 6.1 / PGII
WGK Germany 
3
RTECS 
CG3325000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?