EMAIL THIS PAGE TO A FRIEND
11502 Sigma-Aldrich

Lead

purum, granulated

Synonym: Lead element

Purchase

Properties

grade   purum
quality   granulated
resistivity   20.65 μΩ-cm
bp   1740 °C (lit.)
mp   327.4 °C (lit.)
cation traces   Ag: ≤5 mg/kg
  As: ≤1 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤10 mg/kg
  Sb: ≤50 mg/kg
  Sn: ≤50 mg/kg
SMILES string   [Pb]
InChI   1S/Pb
InChI key   WABPQHHGFIMREM-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
nwg
RTECS 
OF7525000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?