• USA Home
  • 116114 - Trichloroacetic acid

EMAIL THIS PAGE TO A FRIEND
116114 Sigma-Aldrich

Trichloroacetic acid

reagent grade, 98%

Synonym: TCA

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Organic Acids, Synthetic Reagents More...
grade   reagent grade
vapor pressure   1 mmHg ( 51 °C)
  1 mmHg ( 51 °C)
vapor density   <1 (vs air)
assay   98%
refractive index   n20/D 1.62 (lit.)
bp   196 °C (lit.)
mp   54-58 °C (lit.)
solubility   water: soluble 81.7 g/L at 20 °C
density   1.62 g/mL at 25 °C (lit.)
SMILES string   OC(=O)C(Cl)(Cl)Cl
InChI   1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)
InChI key   YNJBWRMUSHSURL-UHFFFAOYSA-N

Description

Application

Traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. Used as decalcifier and fixative in microscopy.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1839 8 / PGII
WGK Germany 
2
RTECS 
AJ7875000
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

T4885 Trichloroacetic acid, ≥99.0% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?