• USA Home
  • 13227 - Manganese(II) lactate trihydrate

EMAIL THIS PAGE TO A FRIEND
13227 Sigma-Aldrich

Manganese(II) lactate trihydrate

puriss., ≥99%

  • CAS Number 6505-50-6

  • Linear Formula (C3H5O3)2Mn · 3H2O

  • Molecular Weight 287.12

  •  Beilstein/REAXYS Number 6118575

  •  EC Number 240-182-0

  •  MDL number MFCD00150481

  •  PubChem Substance ID 24848044

Purchase

Properties

grade   puriss.
assay   ≥99%
impurities   ≤0.002% heavy metals (as Pb)
pH   5-7 (20 °C, 5%)
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   Fe: ≤20 mg/kg
SMILES string   O.O.O.CC(O)C(=O)O[Mn]OC(=O)C(C)O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?