EMAIL THIS PAGE TO A FRIEND
13457 Sigma-Aldrich

Sodium persulfate

≥99%

Synonym: Sodium peroxodisulfate

Purchase

Properties

InChI Key   CHQMHPLRPQMAMX-UHFFFAOYSA-L
assay   ≥99%
impurities   ≤0.003% heavy metals (as Pb)
anion traces   chloride (Cl-): ≤50 mg/kg
cation traces   Fe: ≤20 mg/kg
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 1505 5.1 / PGIII
WGK Germany 
1
RTECS 
SE0525000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?