EMAIL THIS PAGE TO A FRIEND
137133 SAFC

Ethanolamine

EMPROVE® EXPERT, NF

Synonym: 2-Aminoethanol, 2-Aminoethyl alcohol, ETA, MEA 90, MEA-LCI, MEA, Monoethanolamine

  • CAS Number 141-43-5

  • Linear Formula NH2CH2CH2OH

  • Molecular Weight 61.08

  •  Beilstein/REAXYS Number 505944

  •  MDL number MFCD00008183

Purchase

Properties

Quality Level   500
biological source   synthetic
Agency/Method   NF
vapor density   2.1 (vs air)
vapor pressure   0.2 mmHg ( 20 °C)
autoignition temp.   1436 °F
expl. lim.   17 %
packaging   pkg of 1 L (1371331000)
  pkg of 20 L (1371339020)
refractive index   n20/D 1.454 (lit.)
bp   170 °C (lit.)
  69-70 °C/10 mmHg
mp   10-11 °C (lit.)
density   1.012 g/mL at 25 °C (lit.)
SMILES string   NCCO
InChI   1S/C2H7NO/c3-1-2-4/h4H,1-3H2
InChI key   HZAXFHJVJLSVMW-UHFFFAOYSA-N

Description

Linkage

Emprove® Documentation

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Target organs 
Respiratory system
WGK Germany 
WGK 1
Flash Point(F) 
195.8 °F - Pensky-Martens closed cup
Flash Point(C) 
91 °C - Pensky-Martens closed cup

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?