• USA Home
  • 15133 - Boc-D-Met-OH (dicyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
15133 Sigma-Aldrich

Boc-D-Met-OH (dicyclohexylammonium) salt

purum, ≥98.0% (TLC)

Synonym: Boc-D-methionine (dicyclohexylammonium) salt

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Methionine, Peptide Chemistry More...
InChI Key   SKDIPRMGDVUUQO-HMZWWLAASA-N
grade   purum
assay   ≥98.0% (TLC)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?