EMAIL THIS PAGE TO A FRIEND
16179 Sigma-Aldrich

SODOSIL RA 22

acid rinse with citric acid

  •  UBA Number 08050018

Purchase

Properties

Related Categories Cleaners, Detergents, Labware More...
pH   1-2 (20 °C, direct)
density   1.11-1.17 at 20 °C
storage temp.   2-8°C

Description

Application

SODOSIL was used in cleaning plastic ware during determination of lead in whole blood specimens. It was used in rinsing epoxy-coated surfaces, multielectrode arrays and mineral surfaces.

Legal Information

SODOSIL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?