• USA Home
  • 16920 - 4-Bromo-N,N-dimethylaniline

EMAIL THIS PAGE TO A FRIEND
16920 Sigma-Aldrich

4-Bromo-N,N-dimethylaniline

purum, ≥98.0% (T)

  • CAS Number 586-77-6

  • Linear Formula BrC6H4N(CH3)2

  • Molecular Weight 200.08

  •  Beilstein Registry Number 2206155

  •  EC Number 209-582-2

  •  MDL number MFCD00000093

  •  PubChem Substance ID 24850266

Purchase

Properties

InChI Key   XYZWMVYYUIMRIZ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
bp   264 °C(lit.)
mp   52-54 °C(lit.)
  53-55 °C
storage temp.   2-8°C

Description

Other Notes

This product has been replaced by 242950-ALDRICH | 4-Bromo-N,N-dimethylaniline 97%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
BW9300000
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?