• USA Home
  • 17181 - Nutrient Broth pH 6.9 without NaCl

EMAIL THIS PAGE TO A FRIEND
17181 Millipore

Nutrient Broth pH 6.9 without NaCl

for microbiology

  •  NACRES NA.85

Purchase

Properties

Related Categories All Food Control Media (alphabetical), All Liquid Media/Broths, Bacteria (General Media), Clinical Media, Enterobacter,
Quality Level   100
grade   for microbiology
form   powder
shelf life   limited shelf life, expiry date on the label
composition   beef extract, 3 g/L
  peptic digest of animal tissue, 5 g/L
application(s)   microbiological culture: suitable
final pH   6.9±0.2 (25 °C)
Featured Industry   Agriculture
Clinical Testing
Environmental
Food and Beverages
Veterinary
suitability   nonselective for Enterobacter spp.
  nonselective for Enterococcus spp.
  nonselective for Escherichia coli
  nonselective for Salmonella spp.
  nonselective for Staphylococcus spp.
  nonselective for bacteria (General Media)
  nonselective for coliforms

Description

Application

Used as general purpose medium for the cultivation of microorganisms.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the natural environment, microbes have adapted to the habitats most suitable ...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?