• USA Home
  • 17926 - 9-Carbazolepropionic acid

EMAIL THIS PAGE TO A FRIEND
17926 Sigma-Aldrich

9-Carbazolepropionic acid

purum, ≥99.0% (T)

Synonym: 3-(9-Carbazolyl)propionic acid, 9-(2-Carboxyethyl)carbazole

  • CAS Number 6622-54-4

  • Empirical Formula (Hill Notation) C15H13NO2

  • Molecular Weight 239.27

  •  Beilstein Registry Number 203268

  •  MDL number MFCD00085920

  •  PubChem Substance ID 329751583

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Biochemicals and Reagents, Carboxylic Acids and Anhydrides, Derivatization Reagents,
InChI Key   ZAJJJOMMWPVJMH-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (T)
mp   172-176 °C
Gene Information   human ... FABP3(2170), FABP4(2167), FABP5L7(728641)

Description

Other Notes

Reagent for pre-column derivatization of amino acids for fluorescent determination in HPLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?