EMAIL THIS PAGE TO A FRIEND
18118 Supelco

Angelicin

analytical standard

Synonym: 2-Oxo-(2H)-furo(2,3-h)-1-benzopyran, 2H-Furo[2,3-h]-1-benzopyran-2-one, Isopsoralen, NSC 404563

  • CAS Number 523-50-2

  • Empirical Formula (Hill Notation) C11H6O3

  • Molecular Weight 186.16

  •  Beilstein/REAXYS Number 153970

  •  MDL number MFCD00064930

  •  PubChem Substance ID 329751628

  •  NACRES NA.24

Purchase

Properties

Related Categories A - D, Alphabetical Index of Phytochemical Standards, Analytical Standards, Analytical/Chromatography, Angelica,
Quality Level   100
grade   analytical standard
assay   ≥98.0% (HPLC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
impurities   ≤5.0% water
Featured Industry   Food and Beverages
format   neat
storage temp.   2-8°C
SMILES string   O=C1Oc2c(C=C1)ccc3occc23
InChI   1S/C11H6O3/c12-10-4-2-7-1-3-9-8(5-6-13-9)11(7)14-10/h1-6H
InChI key   XDROKJSWHURZGO-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Angular furocoumarin with diverse photobiological effects. Upon long-wavelength UV irradiation, forms monoadduct with double-stranded DNA and reacts with unsaturated fatty acids. Inhibits DNA and RNA synthesis and cell replication in Ehrlich ascites tumor cells.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Precautionary statements 
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
LV0940000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

HPLC Analysis of Furocoumarins on Ascentis® Express RP-Amide

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Furocoumarins on Ascentis® Express RP-Amide
Keywords: Chromatography, High performance liquid chromatography, Vitamins

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?