• USA Home
  • 18245 - 2′-Hydroxyacetophenone

EMAIL THIS PAGE TO A FRIEND
18245 Supelco

2′-Hydroxyacetophenone

analytical standard

Synonym: 2-Acetylphenol

  • CAS Number 118-93-4

  • Linear Formula HOC6H4COCH3

  • Molecular Weight 136.15

  •  Beilstein/REAXYS Number 386123

  •  EC Number 204-288-0

  •  MDL number MFCD00002219

  •  PubChem Substance ID 329751655

  •  NACRES NA.24

Purchase

Properties

Related Categories A-L, Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted, Almond, Analytical Standards,
Quality Level   100
grade   analytical standard
vapor density   4.7 (vs air)
vapor pressure   ~0.2 mmHg ( 20 °C)
assay   ≥97.0% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.558 (lit.)
  n20/D 1.559
bp   213 °C/717 mmHg (lit.)
mp   3-6 °C (lit.)
density   1.133 g/mL at 20 °C
  1.131 g/mL at 25 °C (lit.)
Featured Industry   Cleaning Products
Cosmetics
Flavors and Fragrances
Food and Beverages
Personal Care
format   neat
SMILES string   CC(=O)c1ccccc1O
InChI   1S/C8H8O2/c1-6(9)7-4-2-3-5-8(7)10/h2-5,10H,1H3
InChI key   JECYUBVRTQDVAT-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
RTECS 
AM8575000
Flash Point(F) 
222.8 °F - closed cup
Flash Point(C) 
106 °C - closed cup
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?