EMAIL THIS PAGE TO A FRIEND
210315 Sigma-Aldrich

Acid Red 114

Dye content 45 %

  • CAS Number 6459-94-5

  • Empirical Formula (Hill Notation) C37H28N4Na2O10S3

  • Molecular Weight 830.81

  •  Colour Index Number 23635

  •  EC Number 229-272-0

  •  MDL number MFCD00003894

  •  PubChem Substance ID 24852628

Purchase

Properties

Related Categories A, Cell Biology, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   HNBQFKZSMFFZQY-UHFFFAOYSA-L
composition   Dye content, 45%
λmax   514 nm (2nd)
  -365 nm
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
QJ6475500

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Carcinogen Selector - Digestive System - Large Intestine

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Large Intestine
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Digestive System - Liver

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Liver
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Digestive System - Small Intestine

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Small Intestine
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Digestive System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Clitoral/Preputial Gland

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Endocrine System - Clitoral/Preputial Gland
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Ear Zymbal's Gland

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Liver Endocrine

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Integumentary System - Skin

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Integumentary System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System - Clitoral Gland

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System - Lung

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Reproductive System
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Respiratory System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?