EMAIL THIS PAGE TO A FRIEND
212253 Sigma-Aldrich

Disperse Yellow 9

Dye content 85 %

Synonym: N-(2,4-Dinitrophenyl)-p-phenylenediamine, N-(2,4-Dinitrophenyl)-1,4-phenylenediamine

Purchase

Properties

InChI Key   JMDHCJDATBJFJS-UHFFFAOYSA-N
composition   Dye content, 85%
mp   187-190 °C (lit.)
λmax   359 nm
SMILES string   Nc1ccc(Nc2ccc(cc2[N+]([O-])=O)[N+]([O-])=O)cc1
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?