EMAIL THIS PAGE TO A FRIEND
21277 Sigma-Aldrich

Calpain Inhibitor I

≥97.0% (TLC)

Synonym: ALLN, Ac-LLnL-CHO, MG-101, N-Acetyl-Leu-Leu-Norleu-al, N-Acetyl-L-leucyl-L-leucyl-L-norleucinal

  • CAS Number 110044-82-1

  • Empirical Formula (Hill Notation) C20H37N3O4

  • Molecular Weight 383.53

  •  Beilstein Registry Number 7656053

  •  MDL number MFCD00065505

  •  PubChem Substance ID 329752127

Purchase

Properties

Related Categories Aldehydes, Biochemicals and Reagents, Building Blocks, C13-C60, Carbonyl Compounds,
InChI Key   FMYKJLXRRQTBOR-BZSNNMDCSA-N
assay   ≥97.0% (TLC)
storage temp.   −20°C
Gene Information   human ... CAPN1(823), CTSB(1508), PSMA1(5682)

Description

Other Notes

Shows marked inhibition of cathepsin L

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?