• USA Home
  • 21623 - Antimony(III) chloride reagent (free of CHC)

EMAIL THIS PAGE TO A FRIEND
21623 Supelco

Antimony(III) chloride reagent (free of CHC)

for TLC derivatization, SbCl3 29-31 %

Synonym: Carr-Price Reagent (free of CHC)

  •  NACRES NA.22

Purchase

Description

General description

Antimony(III) chloride is a catalysing agent. The catalytic effect were initially studied in urea, benzaldehyde and methyl isobutyrylacetate to give 6-isopropyl-5-methoxycarbonyl-4-phenyl-3,4-dihydropyrimidin-2(1H)-one.

Application

Antimony(III) chloride may be used as a catalyst in Biginelli reaction of urea, aromatic aldehydes and acetoacetate esters yielding dihydropyrimidinones.

Preparation Note

prepared with Sb2O3 and hydrochloric acid

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3264 8 / PGII
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?