EMAIL THIS PAGE TO A FRIEND
224855 Sigma-Aldrich

Potassium bromate

ReagentPlus®, ≥99%

Purchase

Properties

Related Categories Essential Chemicals, Potassium Salts, Research Essentials, Salts
grade   ReagentPlus®
InChI Key   OCATYIAKPYKMPG-UHFFFAOYSA-M
assay   ≥99%

Description

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1484 5.1 / PGII
WGK Germany 
3
RTECS 
EF8725000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?