• USA Home
  • 24593 - Hamilton® syringe, 7000 series, knurled hub
EMAIL THIS PAGE TO A FRIEND
24593
Hamilton® syringe, 7000 series, knurled hub
7102, volume 2 μL, needle size 23 ga (blunt tip), needle L 70 mm (2.75 in.)
Purchase
Properties
Related Categories 2.0 μL, Analytical/Chromatography, GC Manual Syringes, Hamilton Modified Microliter Syringes, 7000 Series, Liquid sampling,
material   blunt tip stainless steel needle (point style 3)
needle size   23 ga (blunt tip)
feature   Sample contained in needle (no dead volume)
packaging   pkg of 1 ea
mfr. no.   Hamilton, 88500
needle L   70 mm (2.75 in.)
volume   2 μL
Description
General description
• Rapid injection of minute volumes
• Eliminates dead volume by placing the sample in the needle
• Leak-proof at injection pressures up to 6000 psi
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed. A knurled hub is used as it makes a leak-tight seal between the barrel and needle.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15
References
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?