• USA Home
  • 26080 - (±)-3-Chloro-1,2-propanediol

EMAIL THIS PAGE TO A FRIEND
26080 Sigma-Aldrich

(±)-3-Chloro-1,2-propanediol

purum, ≥98.0% (GC)

Synonym: α-Chlorohydrin, α-Glycerol chlorohydrin, α-Monochlorohydrin, 3-MCPD

  • CAS Number 96-24-2

  • Linear Formula ClCH2CH(OH)CH2OH

  • Molecular Weight 110.54

  •  Beilstein/REAXYS Number 635684

  •  EC Number 202-492-4

  •  MDL number MFCD00004712

  •  PubChem Substance ID 329752852

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Building Blocks, Chemical Synthesis, Organic Building Blocks,
vapor pressure   0.04 mmHg ( 25 °C)
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.480 (lit.)
  n20/D 1.480
bp   213 °C (lit.)
density   1.322 g/mL at 25 °C (lit.)
Featured Industry   Agriculture
Environmental
SMILES string   OCC(O)CCl
InChI   1S/C3H7ClO2/c4-1-3(6)2-5/h3,5-6H,1-2H2
InChI key   SSZWWUDQMAHNAQ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2689 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

107271 (±)-3-Chloro-1,2-propanediol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?