EMAIL THIS PAGE TO A FRIEND
27242 Sigma-Aldrich

Trichloroacetic acid

puriss., meets analytical specification of Ph. Eur., USP 21, 99-100.5% (calc. to the dried substance)

Synonym: TCA

  • CAS Number 76-03-9

  • Linear Formula Cl3CCOOH

  • Molecular Weight 163.39

  •  Beilstein/REAXYS Number 970119

  •  EC Number 200-927-2

  •  MDL number MFCD00004177

  •  PubChem Substance ID 57648227

  •  NACRES NA.21

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Organic Acids, Synthetic Reagents More...
Quality Level   200
vapor density   <1 (vs air)
vapor pressure   1 mmHg ( 51 °C)
grade   puriss.
assay   99-100.5% (calc. to the dried substance)
quality   meets analytical specification of Ph. Eur., USP 21
ign. residue   ≤0.02% (as SO4)
loss   ≤0.5% loss on drying, 18 h (on silica gel)
refractive index   n20/D 1.62 (lit.)
bp   196 °C (lit.)
mp   54-58 °C (lit.)
solubility   soluble 81.7 g/L at 20 °C (completely)
density   1.62 g/mL at 25 °C (lit.)
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤500 mg/kg
cation traces   Fe: ≤20 mg/kg
SMILES string   OC(=O)C(Cl)(Cl)Cl
InChI   1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)
InChI key   YNJBWRMUSHSURL-UHFFFAOYSA-N

Description

Application

Traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. Used as decalcifier and fixative in microscopy.

Trichloroacetic acid may be used in catalytic amounts to enhance the yield of benzenediazonium-2-carboxylate from anthranilic acid.

Traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. Used as decalcifier and fixative in microscopy.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Respiratory system
RIDADR 
UN 1839 8 / PGII
WGK Germany 
WGK 2
RTECS 
AJ7875000
Protocols & Articles

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?