EMAIL THIS PAGE TO A FRIEND
27650 Sigma-Aldrich

Colchicine

≥96.0% (HPLC)

Synonym: (S)-N-(5,6,7,9-Tetrahydro-1,2,3,10-tetramethoxy-9-oxobenzo[a]heptalen-7-yl)acetamide

  • CAS Number 64-86-8

  • Empirical Formula (Hill Notation) C22H25NO6

  • Molecular Weight 399.44

  •  Beilstein Registry Number 2228813

  •  EC Number 200-598-5

  •  MDL number MFCD00078484

  •  PubChem Substance ID 329753008

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Inhibitors, Enzyme Inhibitors by Type, Enzymes, Inhibitors, and Substrates, Substrate Analogs More...
InChI Key   IAKHMKGGTNLKSZ-INIZCTEOSA-N
assay   ≥96.0% (HPLC)
impurities   ≤10% ethyl acetate + H2O
  ≤3% water
mp   150-160 °C (dec.)(lit.)

Description

Other Notes

Binds specifically with tubulin thus interfering with microtubule organization.

Price and Availability


Laboratory Gloves

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1544PSN1 6.1 / PGI
WGK Germany 
3
RTECS 
GH0700000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C9754 Colchicine, ≥95% (HPLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?