• USA Home
  • 27666 - Collagen from rat tail

EMAIL THIS PAGE TO A FRIEND
27666 Sigma-Aldrich

Collagen from rat tail

for cell biology, soluble

Purchase

Properties

Related Categories Biochemicals and Reagents, Collagen, Proteins and Derivatives, Structural Proteins
biological source   rat tail
grade   for cell biology
form   powder
quality   soluble
application(s)   cell culture | mammalian: suitable
solubility   H2O: 1 mg/mL, slightly hazy, colorless
suitability   consistent with rat tail collagen for electrophoresis test
storage temp.   2-8°C
Gene Information   rat ... Col1a1(29393)

Description

Application

This product is intended to produce thin layer coatings on tissue culture plates to facilitate attachment of anchorage-dependent cells, recommended for use at 6-10 μg/cm2. It is NOT intended for production of 3-D gels. Type I collagen is often used in cell culture as an attachment substratum with myoblasts, spinal ganglia, hepatocytes, embryonic lung, heart explants, fibroblasts, endothelial cells, and islet cells have all been cultured successfully on films or gels of type I collagen. Collagen type I may also be used in research of Idiopathic pulmonary fibrosis (IPF), studies on the effect of ER stress IPF on lung fibroblasts. Collagen in acidic solution can produce three dimensional scaffolding with use in bioengineering and cell culture applications.

Soluble rat-tail tendon cartilage from normal and experimentally-induced diabetic animals were compared in an attempt to elucidate the reasons for the altered response of diabetic connective tissues to inflammation and trauma. The collagen from diabetic animals was markedly more viscous and less mobile on electrophoresis, suggesting higher levels of polymerization.

Biochem/physiol Actions

Type I collagen is a component of skin, bone, tendon, and other fibrous connective tissues.

Components

All collagen molecules are composed of three polypeptide chains arranged in a triple helical conformation, with a primary structure that is mostly a repeating motif with glycine in every third position and proline or 4-hydroxyproline frequently preceding the glycine residue. Type I collagen differs from other collagens by its low lysine hydroxylation and low carbohydrate composition.

Preparation Note

This product was prepared by a modification of the extraction method of Bornstein, M.B., Lab. Invest., 7, 134 (1958). It is soluble in water at 1 mg/mL, producing a slightly hazy, colorless solution.

Other Notes

Collagen is classified into a number of structurally and genetically distinct types. We use the nomenclature proposed by Bornstein and Traub. Do not confuse Sigma type designations with recognized collagen classification types.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?