• USA Home
  • 277266 - Luxol® Brilliant Green BL

EMAIL THIS PAGE TO A FRIEND
277266 Sigma-Aldrich

Luxol® Brilliant Green BL

Complex of 1,3-di-o-tolylguanidine and Acid Green 3 (2:1)

Synonym: Solvent Green 11

  • CAS Number 71799-14-9

  • Empirical Formula (Hill Notation) C52H52N5NaO6S2

  • Molecular Weight 930.12

  •  MDL number MFCD00064439

  •  PubChem Substance ID 24856747

Purchase

Properties

absorption   λmax 624 nm

Description

Legal Information

Luxol is a registered trademark of Rohm and Haas Co.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?