EMAIL THIS PAGE TO A FRIEND
284882 Sigma-Aldrich

Pinacryptol yellow

Dye content 98 %

Synonym: 6-Ethoxy-1-methyl-2-(3-nitro-β-styryl)quinolinium methyl sulfate

  • CAS Number 25910-85-4

  • Empirical Formula (Hill Notation) C21H22N2O7S

  • Molecular Weight 446.47

  •  Beilstein/REAXYS Number 3881592

  •  EC Number 247-336-6

  •  MDL number MFCD00011967

  •  PubChem Substance ID 24857182

Purchase

Properties

InChI Key   ZXQHSPWBYMLHLB-BXTVWIJMSA-M
composition   Dye content, 98%
mp   260-262 °C (lit.)
λmax   385 nm
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?