• USA Home
  • 28628-U - Hamilton® 1005LTN syringe

EMAIL THIS PAGE TO A FRIEND
28628-U

Hamilton® 1005LTN syringe

1005LTN, volume 5 mL, needle size 23 ga (cone tip, side port), needle L 56 mm (2.2 in.)

  •  NACRES NB.03

Purchase

Properties

Related Categories Analytical/Chromatography, CTC/Leap Technologies Autosamplers, GC Autosampler Syringes, GC Headspace Sampling: CTC Combi PAL and GC PAL, Syringes and Syringe Accessories More...
material   cone tip, side port stainless steel (point style 5)
needle size   23 ga (cone tip, side port)
description   gastight
feature   GASTIGHT® Syringe, Fixed Needle
packaging   pkg of 1 ea
mfr. no.   Hamilton®, 203086
needle L   56 mm (2.2 in.)
volume   5 mL
compatibility   for use with CTC COMBI Pal (luer tip needle)

Description

Legal Information

GASTIGHT is a registered trademark of Hamilton Co.

Hamilton is a registered trademark of Hamilton Co.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?