• USA Home
  • 28995 - cis-1,2-Cyclohexanediol

EMAIL THIS PAGE TO A FRIEND
28995 Sigma-Aldrich

cis-1,2-Cyclohexanediol

puriss., ≥99.0% (GC)

  • CAS Number 1792-81-0

  • Linear Formula C6H10(OH)2

  • Molecular Weight 116.16

  •  Beilstein Registry Number 1340578

  •  MDL number MFCD00064944

  •  PubChem Substance ID 329753304

Purchase

Properties

InChI Key   PFURGBBHAOXLIO-OLQVQODUSA-N
grade   puriss.
assay   ≥99.0% (GC)
mp   97-101 °C(lit.)
  97-101 °C

Description

Other Notes

Starting material for several enantioselective enzymatic transformations, e.g. mono-acylation; mono-oxidation

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

361267 cis-1,2-Cyclohexanediol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?