EMAIL THIS PAGE TO A FRIEND
29740 Sigma-Aldrich

Cyclopentanol

purum, ≥99.0% (GC)

  • CAS Number 96-41-3

  • Linear Formula C5H9OH

  • Molecular Weight 86.13

  •  Beilstein Registry Number 1900556

  •  EC Number 202-504-8

  •  MDL number MFCD00001363

  •  PubChem Substance ID 24857911

Purchase

Properties

InChI Key   XCIXKGXIYUWCLL-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.453(lit.)
  n20/D 1.453
bp   139-140 °C(lit.)
mp   −19 °C(lit.)
density   0.948 g/mL at 20 °C
  0.949 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 2244 3 / PGIII
WGK Germany 
1
Flash Point(F) 
123.8 °F
Flash Point(C) 
51 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

C112208 Cyclopentanol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?