• USA Home
  • 302619 - Azure B tetrafluoroborate

EMAIL THIS PAGE TO A FRIEND
302619 Sigma-Aldrich

Azure B tetrafluoroborate

Dye content 95 %

Purchase

Properties

Related Categories A, Cell Biology, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   OYVZJZSEBPXXFF-UHFFFAOYSA-O
composition   Dye content, 95%
mp   244 °C (dec.)(lit.)
absorption   λmax 637 nm

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?


Tissue Diagnostics

Milli-Q® Water Purification Solutions

Biomedical Applications