EMAIL THIS PAGE TO A FRIEND
30642 Supelco

2-Acetylpyrrole

analytical standard

Synonym: Methyl 2-pyrrolyl ketone

  • CAS Number 1072-83-9

  • Empirical Formula (Hill Notation) C6H7NO

  • Molecular Weight 109.13

  •  Beilstein/REAXYS Number 1882

  •  EC Number 214-016-2

  •  MDL number MFCD00005220

  •  PubChem Substance ID 329753554

  •  NACRES NA.24

Purchase

Properties

Related Categories A-F, A-L, Additional Standards, Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted,
Quality Level   100
grade   analytical standard
assay   ≥98.5% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
impurities   ≤0.5% water
bp   220 °C (lit.)
mp   88-93 °C (lit.)
Featured Industry   Cleaning Products
Cosmetics
Flavors and Fragrances
Food and Beverages
Personal Care
format   neat
SMILES string   CC(=O)c1ccc[nH]1
InChI   1S/C6H7NO/c1-5(8)6-3-2-4-7-6/h2-4,7H,1H3
InChI key   IGJQUJNPMOYEJY-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
OB5970000
Protocols & Articles

Articles

GC Analysis of Volatiles in Roasted Coffee Beans on Omegawax® after SPME using a 50/30 μm DVB/Carboxen/PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Volatiles in Roasted Coffee Beans on Omegawax® after SPME using a 50/30 μm DVB/Carboxen/PDMS Fiber
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Size-exclusion chromatography, Solid phase microextractions

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?