• USA Home
  • 31066 - Silver sulfate-Sulfuric acid solution

EMAIL THIS PAGE TO A FRIEND
31066 Supelco

Silver sulfate-Sulfuric acid solution

for COD determination according to DIN 38409, part 41, solution (volumetric)

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Catalysis and Inorganic Chemistry, Chemical Synthesis, Potable- & Waste-Water Testing,
Quality Level   100
grade   for COD determination according to DIN 38409, part 41
form   solution (volumetric)
reaction suitability   reagent type: catalyst
core: silver
concentration   60 g/L in sulfuric acid
Featured Industry   Environmental
General Analytical
storage temp.   room temp
SMILES string   [Ag+].[Ag+].[O-]S([O-])(=O)=O
InChI   1S/2Ag.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2
InChI key   YPNVIBVEFVRZPJ-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1830 8 / PGII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?