EMAIL THIS PAGE TO A FRIEND
31206 Supelco

Atraton

PESTANAL®, analytical standard

Synonym: Atrazine-methoxy

  • CAS Number 1610-17-9

  • Empirical Formula (Hill Notation) C9H17N5O

  • Molecular Weight 211.26

  •  Beilstein/REAXYS Number 613098

  •  EC Number 216-547-5

  •  MDL number MFCD00078647

  •  PubChem Substance ID 329753677

  •  NACRES NA.24

Purchase

Properties

Related Categories A-B, A-D, Additional Standards, All Herbicide Standards, Alpha Index of All Pesticide Standards,
Quality Level   100
grade   analytical standard
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat
storage temp.   room temp
SMILES string   CCNc1nc(NC(C)C)nc(OC)n1
InChI   1S/C9H17N5O/c1-5-10-7-12-8(11-6(2)3)14-9(13-7)15-4/h6H,5H2,1-4H3,(H2,10,11,12,13,14)
InChI key   PXWUKZGIHQRDHL-UHFFFAOYSA-N

Description

General description

Atraton is a triazine herbicide intended to control weeds in a variety of crops. Triazines herbicides act by inhibiting photosynthesis in all organisms with oxygen-evolving photosystems.

Application

Atraton may be used as a reference standard for the determination of the analyte:
• In environmental water samples using liquid chromatography-electrospray ionization mass spectrometry (LC/ESI/MS).
• In fruits using an ultrasonic-assisted matrix solid-phase dispersion (UA-MSPD) method.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
XY8750000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?