EMAIL THIS PAGE TO A FRIEND
31484 Supelco

Binapacryl

PESTANAL®, analytical standard

  • CAS Number 485-31-4

  • Empirical Formula (Hill Notation) C15H18N2O6

  • Molecular Weight 322.31

  •  Beilstein/REAXYS Number 2013712

  •  EC Number 207-612-9

  •  MDL number MFCD00055425

  •  PubChem Substance ID 329753784

  •  NACRES NA.24

Purchase

Properties

Related Categories Acaricides, Analytical Standards, Analytical/Chromatography, Food and Cosmetic Component Standards, Pesticides & Metabolites Standards,
Quality Level   100
grade   analytical standard
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Cleaning Products
Cosmetics
Environmental
Food and Beverages
Personal Care
format   neat
storage temp.   room temp
SMILES string   CCC(C)c1cc(cc(c1OC(=O)\C=C(/C)C)[N+]([O-])=O)[N+]([O-])=O
InChI   1S/C15H18N2O6/c1-5-10(4)12-7-11(16(19)20)8-13(17(21)22)15(12)23-14(18)6-9(2)3/h6-8,10H,5H2,1-4H3
InChI key   ZRDUSMYWDRPZRM-UHFFFAOYSA-N

Description

General description

Binapacryl belongs to the dinitrophenol family of chemicals primarily used as a fungicide against powdery mildews on apples, cucumbers and melons.

Application

Binapacryl may be used as a reference standard for the determination of the analyte in apple and cucumber samples by high-performance liquid chromatography method.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN2811 - class 6.1 - PG 3 - SMP - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
WGK 3
RTECS 
GQ5600000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?