EMAIL THIS PAGE TO A FRIEND
318841 Sigma-Aldrich

Cresol red solution

0.04 wt. % in H2O

  • CAS Number 1733-12-6

  • Empirical Formula (Hill Notation) C21H18O5S

  • Molecular Weight 382.43

  •  Beilstein/REAXYS Number 343399

  •  MDL number MFCD00001618

Purchase

Properties

concentration   0.04 wt. % in H2O
pH range   0.0 - 1.0, red to yellow
density   0.98 g/mL at 25 °C
λmax   425 nm
SMILES string   [Na+].Cc1cc(ccc1O)\C(=C2/C=CC(=O)C(C)=C2)c3ccccc3S([O-])(=O)=O
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1170 - class 3 - PG 3 - Ethanol, solution
WGK Germany 
1
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?