• USA Home
  • 32145 - Tolfenamic acid-(benzoic ring-13C6)

EMAIL THIS PAGE TO A FRIEND
32145 Supelco

Tolfenamic acid-(benzoic ring-13C6)

VETRANAL, analytical standard

Synonym: 2-(3-Chloro-2-methylphenylamino)benzoic-13C6 acid

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Chromatography, Drugs & Metabolites,
grade   VETRANAL, analytical standard
InChI Key   YEZNLOUZAIOMLT-CDLMHYHCSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?