• USA Home
  • 32313 - Potassium oxalate monohydrate

EMAIL THIS PAGE TO A FRIEND
32313 Sigma-Aldrich

Potassium oxalate monohydrate

puriss. p.a., ACS reagent, 99.5-101.0%

Synonym: Ethanedioic acid, Oxalic acid potassium salt

  • CAS Number 6487-48-5

  • Linear Formula (COOK)2 · H2O

  • Molecular Weight 184.23

  •  Beilstein/REAXYS Number 3752576

  •  EC Number 209-506-8

  •  MDL number MFCD00150033

  •  PubChem Substance ID 57648646

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Essential Chemicals, O-P, Puriss p.a. ACS,
grade   ACS reagent
  puriss. p.a.
assay   99.5-101.0%
impurities   ≤0.001% heavy metals (as Pb)
  ≤0.001% total nitrogen (N)
pH   7.0-8.5 (20 °C, 5%)
anion traces   chloride (Cl-): ≤10 mg/kg
  phosphate (PO43-): ≤10 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   Fe: ≤10 mg/kg
  Na: ≤200 mg/kg
suitability   passes test for reaction against H2SO4
SMILES string   O.[K+].[K+].[O-]C(=O)C([O-])=O
InChI   1S/C2H2O4.2K.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);;;1H2/q;2*+1;/p-2
InChI key   QCPTVXCMROGZOL-UHFFFAOYSA-L

Description

General description

Potassium oxalate monohydrate (POM) is a potassium salt of oxalic acid. Its crystal structure has been investigated by three-dimensional neutron diffraction studies.

Application

Potassium oxalate monohydrate may be used in the preparation of SBA-15 (Santa Barbara Amorphous-15)- catalyst.

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Single- & multi-channel pipettes from BRAND
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?