• USA Home
  • 32387 - (±)-Propranolol-(isopropyl-d7) hydrochloride

EMAIL THIS PAGE TO A FRIEND
32387 Supelco

(±)-Propranolol-(isopropyl-d7) hydrochloride

VETRANAL, analytical standard

Synonym: (±)-1-Isopropyl-d7-amino-3-(1-naphthyloxy)-2-propanol hydrochloride

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Chromatography, Drugs & Metabolites,
grade   VETRANAL, analytical standard
InChI Key   ZMRUPTIKESYGQW-ODLOEXKQSA-N
storage temp.   2-8°C

Description

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?