• USA Home
  • 32423 - Benazepril hydrochloride

EMAIL THIS PAGE TO A FRIEND
32423 Supelco

Benazepril hydrochloride

VETRANAL®, analytical standard

Synonym: (3S)-3-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid hydrochloride

  • CAS Number 86541-74-4

  • Empirical Formula (Hill Notation) C24H28N2O5 · HCl

  • Molecular Weight 460.95

  •  MDL number MFCD00895734

  •  PubChem Substance ID 329754201

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Antibiotics Standards, Pharmaceuticals, Illicit Drugs & Alcohol
grade   analytical standard
product line   VETRANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Forensics and Toxicology
Pharmaceutical (small molecule)
format   neat
SMILES string   Cl[H].CCOC(=O)[C@H](CCc1ccccc1)N[C@H]2CCc3ccccc3N(CC(O)=O)C2=O
InChI   1S/C24H28N2O5.ClH/c1-2-31-24(30)20(14-12-17-8-4-3-5-9-17)25-19-15-13-18-10-6-7-11-21(18)26(23(19)29)16-22(27)28;/h3-11,19-20,25H,2,12-16H2,1H3,(H,27,28);1H/t19-,20-;/m0./s1
InChI key   VPSRQEHTHIMDQM-FKLPMGAJSA-N
Gene Information   human ... ACE(1636)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Benazepril is a long-acting angiotensin converting enzyme (ACE) inhibitor.

Legal Information

VETRANAL is a registered trademark of Sigma-Aldrich Chemie GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
CX7065000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?