• USA Home
  • 32480 - Clenpenterol-(dimethyl-d6, propyl-d5) hydrochloride

EMAIL THIS PAGE TO A FRIEND
32480 Supelco

Clenpenterol-(dimethyl-d6, propyl-d5) hydrochloride

VETRANAL®, analytical standard

Synonym: 1-(4-Amino-3,5-dichlorophenyl)-2-(1,1-dimethyl-d6-propyl-d5-amino)ethanol hydrochloride

  • CAS Number 1325559-11-2

  • Empirical Formula (Hill Notation) C13D11H9Cl2N2O · HCl

  • Molecular Weight 338.75

  •  MDL number MFCD19704812

  •  PubChem Substance ID 329754238

  •  NACRES NA.24

Purchase

Properties

Related Categories Amphetamines, Analytical Standards, Analytical/Chromatography, Pharmaceuticals, Illicit Drugs & Alcohol
Quality Level   100
grade   analytical standard
product line   VETRANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Forensics and Toxicology
Pharmaceutical (small molecule)
format   neat
mass shift   M+11
SMILES string   Cl.[2H]C([2H])([2H])C([2H])([2H])C(NCC(O)c1cc(Cl)c(N)c(Cl)c1)(C([2H])([2H])[2H])C([2H])([2H])[2H]
InChI   1S/C13H20Cl2N2O.ClH/c1-4-13(2,3)17-7-11(18)8-5-9(14)12(16)10(15)6-8;/h5-6,11,17-18H,4,7,16H2,1-3H3;1H/i1D3,2D3,3D3,4D2;
InChI key   HJGDYFOIRCZBLH-IJXFQFNHSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Sigma-Aldrich Chemie GmbH

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?